

Hydrochloric acid 36.5 - 38%, GenAR® NF, Macron Fine Chemicals™
Catalog # 2515-46
Supplier: MACRON AVANTOR BRAND
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle, clear, Mall, PVC-coated
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Meets J.P. Chemical Specifications:—
- Meets B.P. Chemical Specifications:—
- Meets E.P. Chemical Specifications:—
- Meets N.F. Requirements:—
- GMP Manufactured Product:—
- Store below 25°C:—
- CAUTION: For Manufacturing, processing or repackaging:—
- Bulk Pharmaceutical Chemical:Passes Test
- NF - Identification:Passes Test
- NF - Residue on Ignition:≤ 0.008 %
- NF - Bromide or Iodide:Passes Test
- NF - Free Bromine or Chlorine:Passes Test
- NF - Sulfate (SO₄):Passes Test
- NF - Sulfite (SO₃):Passes Test
- NF - Assay (HCl)(NF):36.5 - 38.0 %
- Identification A (EP/BP):Passes Test
- Identification B (EP/BP):Passes Test
- Identification C (EP/BP):Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Free Chlorine (as Cl):≤ 4 ppm
- EP/BP - Sulfate (SO₄):≤ 20 ppm
- EP/BP - Residue after Evaporation:≤ 0.01 %
- Assay (HCl)(EP/BP):35.0 - 39.0 %
- JP - Assay (HCl)(JP):35.0 - 38.0 %
- JP - Identification:Passes Test
- JP - Residue on ignition, mg/10mL:≤ 1.0
- JP - Bromide or Iodide:Passes Test
- JP - Free Bromine or Chlorine:Passes Test
- JP - Sulfate (SO₄):Passes Test
- JP - Sulfite (SO₃):Passes Test
- JP - Arsenic (As):≤ 1 ppm
- JP - Heavy Metals (as Pb) (JP):≤ 5 ppm
- JP - Mercury (Hg):≤ 0.04 ppm
Specification Test Results
About this item
:
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–30 °C
Density:1.18 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1789
ADR:8,II
:Management of Change (MOC) category = R
:36.46 g/mol
EINECS:231-595-7
:GHS05
:GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324