Order Entry
Italy
ContactUsLinkComponent
 
Ammonium sulphate 38% (w/w) in aqueous solution, BAKER ANALYZED®, J.T.Baker®
Catalog # 0791-09
Supplier:  Avantor
CAS Number:  
undefined
Ammonium sulphate 38% (w/w) in aqueous solution, BAKER ANALYZED®, J.T.Baker®
Catalog # 0791-09
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    PE drum, blue
  • Pk:
    525 lb
  • Seq:
    7332

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    37.0 - 39.0%
  • Assay ((NH₄)₂SO₄) (by formol method):
    37.0 - 39.0 %
  • Color (APHA):
    ≤10
  • pH (As Is" basis)":
    5.7 - 6.3
  • Identification A:
    Passes Test
  • Identification B:
    Passes Test
  • Insoluble Matter:
    ≤0.005 %
  • Residue after Ignition:
    ≤0.005 %
  • Nitrate (NO₃):
    ≤0.001 %
  • Chloride (Cl):
    ≤0.0005 %
  • Phosphate (PO₄):
    ≤0.0005 %
  • Arsenic (As):
    ≤0.00002 %
  • Heavy Metals (as Pb):
    ≤0.0005 %
  • Iron (Fe):
    ≤0.0005 %

Specification Test Results

About this item

 :
 :132.14 g/mol
Density:1.77 g/cm³
CAS Number:7783-20-2
Formula:(NH₄)₂SO₄
MDL Number:MFCD00003391