Order Entry
Italy
ContactUsLinkComponent
Organic reference standards, fluticasones, Ph. Eur.
Organic reference standards, fluticasones, Ph. Eur.
Supplier:  EDQM
Certificates
Organic reference standards, fluticasones, Ph. Eur.
Supplier:  EDQM

Specifications

  • CAS No.:
    73205-13-7
  • Formula:
    C₂₅H₃₂F₂O₅S
  • Description:
    Fluticasone impurity D, 6α,9-difluoro-11β-hydroxy-16α-methyl-17-[(methylsulfanyl)carbonyl]-3-oxoandrosta-1,4-dien-17α-ylpropanoate

Specifications

Frequently Bought Together