Organic reference standards, cisplatins, Ph. Eur.
Supplier: EDQM
Certificates
About this item
Specifications
- CAS No.:14913-33-8
- Formula:Cl₂H₆N₂Pt
- Description:Cisplatin impurity A, trans-diamminedichloroplatinum(II) (transplatin), Ph. Eur. standard
Specifications
Frequently Bought Together