Order Entry
Italy
ContactUsLinkComponent
Anti-p44/42 ERK1/2 Mouse Monoclonal Antibody (KLH (Keyhole Limpet Hemocyanin)) [clone: 2E8-2F6-C12]
Anti-p44/42 ERK1/2 Mouse Monoclonal Antibody (KLH (Keyhole Limpet Hemocyanin)) [clone: 2E8-2F6-C12]
Catalog # ABNOMAB15284
Supplier:  Abnova
Anti-p44/42 ERK1/2 Mouse Monoclonal Antibody (KLH (Keyhole Limpet Hemocyanin)) [clone: 2E8-2F6-C12]
Catalog # ABNOMAB15284
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    p44/42 ERK1/2
  • Clonality:
    Monoclonal
  • Clone:
    2E8-2F6-C12
  • Conjugation:
    KLH (Keyhole Limpet Hemocyanin)
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Modification:
    phospho T202/Y204
  • Storage buffer:
    In PBS (50% glycerol, 0.5% BSA, 0.02% sodium azide).
  • Storage temperature:
    Store at -20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding T202/T204 of human p44/42 ERK1/2.
  • Purification:
    Affinity purification
  • Size:
    100 µL
  • Pk:
    100 µl

Specifications

About this item

Mouse monoclonal antibody raised against synthetic phosphopeptide of human p44/42 ERK1/2.

Type: Primary
Antigen: p44/42 ERK1/2
Clonality: Monoclonal
Clone: 2E8-2F6-C12
Conjugation: KLH (Keyhole Limpet Hemocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse