Order Entry
Italy
ContactUsLinkComponent
Anti-KRT8 Mouse Monoclonal Antibody [clone: C-51]
Anti-KRT8 Mouse Monoclonal Antibody [clone: C-51]
Catalog # ABNOMAB14663
Supplier:  Abnova
Anti-KRT8 Mouse Monoclonal Antibody [clone: C-51]
Catalog # ABNOMAB14663
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cytoskeleton preparation from HeLa cells.
  • Antigen symbol:
    KRT8
  • Clonality:
    Monoclonal
  • Clone:
    C-51
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    3856|3875
  • Immunogen:
    Cytoskeleton preparation from HeLa cells.
  • Purification:
    Protein G purification
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal antibody raised against native human KRT8 and KRT18.

Type: Primary
Antigen: KRT8
Clonality: Monoclonal
Clone: C-51
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human