Order Entry
Italy
ContactUsLinkComponent
Anti-HLA-DPB1 Mouse Monoclonal Antibody [clone: CR3/43]
Anti-HLA-DPB1 Mouse Monoclonal Antibody [clone: CR3/43]
Catalog # ABNOMAB14682
Supplier:  Abnova
Anti-HLA-DPB1 Mouse Monoclonal Antibody [clone: CR3/43]
Catalog # ABNOMAB14682
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    HLA-DPB1
  • Clonality:
    Monoclonal
  • Clone:
    CR3/43
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    3115|3117|3122
  • Storage buffer:
    In 10 mM PBS.
  • Storage temperature:
    Store at 4 °C. For long term storage store at -20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Cells from human tonsil.
  • Purification:
    Protein A/G purification
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal antibody raised against human HLA-DP/HLA-DQ/HLA-DR.

Type: Primary
Antigen: HLA-DPB1
Clonality: Monoclonal
Clone: CR3/43
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human