Order Entry
Italy
ContactUsLinkComponent
 
Anti-CD81 Mouse Monoclonal Antibody (FITC) [clone: MEM-38]
  ABNOMAB15370
 :  Abnova
Anti-CD81 Mouse Monoclonal Antibody (FITC) [clone: MEM-38]
  ABNOMAB15370
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    CD81 molecule
  • Antigen symbol:
    CD81
  • Clonality:
    Monoclonal
  • Clone:
    MEM-38
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    975
  • Storage buffer:
    In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
  • Storage temperature:
    Store in the dark at 4 °C.
  • Immunogen:
    MOLT-4 (human T-ALL cell line).
  • Purification:
    Affinity purification
  • Size:
    100 AS
  • Pk:
    100 Assays

 

 

Mouse monoclonal antibody raised against human CD81.

Type: Primary
Antigen: CD81
Clonality: Monoclonal
Clone: MEM-38
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human