Anti-CD8a Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: 53-6.72]
Catalog # ABNOMAB15306
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:CD8a molecule
- Antigen symbol:CD8a
- Clonality:Monoclonal
- Clone:53-6.72
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
- Host:Rat
- Isotype:IgG2b
- Reactivity:Mouse
- Form:Liquid
- Gene ID:12525
- Storage buffer:In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
- Storage temperature:Store in the dark at 4°C.
- Immunogen:Mouse thymocytes/splenocytes.
- Purification:Affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rat monoclonal antibody raised against mouse Cd8a.
Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: 53-6.72
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse