Anti-ZNRF4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-9142R-A555
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Zinc And Ring Finger 4
- Antigen symbol:ZNRF4
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:148066
- Antigen synonyms:RING finger protein 204|SPERIZIN|ZNRF4_HUMAN.|zinc and ring finger 4|RNF204|Ssrzf1|Zinc/RING finger protein 4 precursor|spzn
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
ZNRF4 contains 1 PA (protease associated) domain and 1 RING-type zinc finger.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: ZNRF4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat