Order Entry
Italy
ContactUsLinkComponent
Anti-CDCA7 Rabbit Polyclonal Antibody (Cy7®)
Anti-CDCA7 Rabbit Polyclonal Antibody (Cy7®)
  BOSSBS-9128R-CY7
 :  Bioss
Anti-CDCA7 Rabbit Polyclonal Antibody (Cy7®)
  BOSSBS-9128R-CY7
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Cell Division Cycle associated 7
  • Antigen symbol:
    CDCA7
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy7®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    83879
  • Antigen synonyms:
    Cell Division Cycle associated protein 7|Cell Division Cycle-associated protein 7|JPO1|c-Myc target JPO1
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

CDCA7 is a nuclear protein without a known function, although its high homology with the transcription factor JPO2 or RAM2 suggests that it is also a transcription factor. CDCA7 is one of many target genes regulated by the c-Myc transcription factor. Overexpression of CDCA7 occurs in a significant proportion of cancers and it may play an important role in tumorigenesis. Normally it is periodically expressed in the cell cycle, peaking at the G1 to S phase transition.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CDCA7
Clonality: Polyclonal
Clone: 0
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: