Order Entry
Italy
ContactUsLinkComponent
Anti-RTFDC1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anti-RTFDC1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
  BOSSBS-9700R-A647
 :  Bioss
Anti-RTFDC1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
  BOSSBS-9700R-A647
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    replication termination factor 2 domain containing 1
  • Antigen symbol:
    RTFDC1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 647
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    51507
  • Antigen synonyms:
    CDAO5|HSPC164|C20orf43|SHUJUN-3
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Representing about 2% of human DNA, chromosome 20 consists of approximately 63 million bases and 600 genes. Chromosome 20 contains a region with numerous genes expressed in the epididymis, which are thought important for seminal production, and some viewed as potential targets for male contraception. The PRNP gene encoding the prion protein associated with spongiform encephalopathies, like Creutzfeldt-Jakob disease, is found on chromosome 20. Amyotrophic lateral sclerosis, spinal muscular atrophy, ring chromosome 20 epilepsy syndrome and Alagille syndrome are also associated with chromosome 20. The C20orf43 gene product has been provisionally designated C20orf43 pending further characterization.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RTFDC1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat