Order Entry
Italy
ContactUsLinkComponent
Anti-PPAP2C Rabbit Polyclonal Antibody (Cy7®)
Anti-PPAP2C Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-8527R-CY7
Supplier:  Bioss
Anti-PPAP2C Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-8527R-CY7
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Phosphatidic Acid Phosphatase Type 2C
  • Antigen symbol:
    PPAP2C
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy7®
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    8612
  • Antigen synonyms:
    PAP2-gamma|PAP2-G|Lipid phosphate phosphohydrolase 2|Phosphatidate phosphohydrolase type 2c|PAP2c|Phosphatidic acid phosphatase 2c|PAP-2c
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

The Vang family of proteins are integral membrane proteins that are homologues of the Drosophila tissue polarity gene strabismus. The gene encoding for Van Gogh-like protein 1 (Vangl1), also designated Strabismus 2 (STB2), localizes to human chromosome 1p11-p13.1. Van Gogh-like protein 2 (Vangl2), also designated Strabismus 1 (STB1), localizes to chromosome 1q22-q23. Vangl1 is expressed primarily in testis and ovary, but is also expressed in gastric and pancreatic cancer. Vangl proteins play a key developmental role in establishing planar cell polarity (PCP) and in regulating convergent extension (CE) movements during embryogenesis. Vangl1 and Vangl2 are both downregulated in several cancer cell lines and primary tumors.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: PPAP2C
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat