Specifications
- Antibody type:Primary
- Antigen name:Nuclear interacting partner of ALK
- Antigen symbol:NIPA
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:51530
- Antigen synonyms:Nuclear interacting partner of anaplastic lymphoma kinase|Zinc finger C3HC type containing 1|Nuclear interacting partner of ALK|ZC3HC1|hNIPA|NIPA_HUMAN.
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
The regulated oscillation of protein expression is an essential mechanism of cell cycle control. The SCF class of E3 ubiquitin ligases is involved in this process by targeting cell cycle regulatory proteins for degradation by the proteasome, with the F-box subunit of the SCF specifically recruiting a given substrate to the SCF core. NIPA (nuclear interaction partner of ALK) is a human F-box-containing protein that defines an SCF-type E3 ligase (SCFNIPA) controlling mitotic entry. Assembly of this SCF complex is regulated by cell-cycle-dependent phosphorylation of NIPA, which restricts substrate ubiquitination activity to interphase. Nuclear cyclin B1 is a substrate of SCFNIPA. Inactivation of NIPA by RNAi results in nuclear accumulation of cyclin B1 in interphase, activation of cyclin B1-Cdk1 kinase activity, and premature mitotic entry. Thus, SCFNIPA-based ubiquitination may regulate S-phase completion and mitotic entry in the mammalian cell cycle.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NIPA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat