- Antibody type:Primary
- Antigen name:Lysine (K)-specific Demethylase 2A
- Antigen symbol:KDM2A
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:22992
- Antigen synonyms:LILINA|CXXC8|FBXL11|JHDM1A|FBL11|FBL7
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
F-box proteins are critical components of the SCF (Skp1-CUL-1-F-box protein) type E3 ubiquitin ligase complex and are involved in substrate recognition and protein recruitment. F-box proteins are members of a large family that regulate the cell cycle, immune response, signaling cascades and developmental programs by targeting proteins, such as cyclins, for degradation by the proteasome after ubiquitination. FBL11, also known as FBXL11 (F-box and leucine-rich repeat protein 11), CXXC8, KDM2A, JHDM1A (JmjC domain-containing histone demethylation protein 1A) or LILINA, is a 1,162 amino acid member of the F-box protein family that contains one F-box domain and localizes to the nucleus. Expressed ubiquitously with highest expression in testis, ovary and brain, FBL11 functions to demethylate the Lys-36 residue of histone H3, thereby modulating the histone code. Additionally, FBL11 is thought to promote the ubiquitination and subsequent degradation of various phosphorylated proteins. Three isoforms of FBL11 exist due to alternative splicing events.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: KDM2A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat