Order Entry
Italy
ContactUsLinkComponent
Anti-PDCD7 Rabbit Polyclonal Antibody (Cy3®)
Anti-PDCD7 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-6834R-CY3
Supplier:  Bioss
Anti-PDCD7 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-6834R-CY3
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Programmed cell death protein 7
  • Antigen symbol:
    PDCD7
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy3®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    10081
  • Antigen synonyms:
    PDCD7_HUMAN|Programmed cell death protein 7.|Pdcd7|HES18|Programmed cell death 7|MGC22015|Apoptosis-related protein ES18|ES18
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

This gene encodes a 59 kDa protein that is associated with the U11 small nuclear ribonucleoprotein (snRNP), which is a component of the minor U12-type spliceosome responsible for catalyzing pre-mRNA splicing of U12-type introns. [provided by RefSeq, Dec 2010].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PDCD7
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat