Anti-RAB25 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
BOSSBS-6501R-FITC
: Bioss
- Antibody type:Primary
- Antigen name:Ras-related protein Rab-25
- Antigen symbol:RAB25
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:57111
- Antigen synonyms:PPP1R93|GP2B|GTA|CD41B|HPA3|GPIIb|BDPLT2|CD41|BDPLT16|GT
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
RAB proteins, such as RAB25, are members of the RASsuperfamily of small GTPases that are involved in membranetrafficking. Members of the RAB11 subfamily, including RAB25,control the return of internalized membrane-associated moieties tothe cell surface
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RAB25
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat