Anti-C7ORF69 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
BOSSBS-5101R-FITC
: Bioss
- Antibody type:Primary
- Antigen name:Chromosome 7 open reading frame 69
- Antigen symbol:C7ORF69
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:80099
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
MFAP3L is a membrane protein that is highly expressed in testis. It contains 1 Ig-like C2-type (immunoglobulin-like) domain and has three named isoforms produced by alternative splicing. Its function remains unknown.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: C7ORF69
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat