Specifications
- Antibody type:Primary
- Antigen name:Adducin 1 Alpha
- Antigen symbol:ADD1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:118
- Antigen synonyms:ADDA|ADD2|Adducin-alpha|alpha-Adducin|Adducin1|Adducin-1|Erythrocyte adducin subunit beta|beta-Adducin|beta Adducin|Beta-adducin|Adducin beta|ADD1|Erythrocyte adducin subunit alpha|Adducin-beta|alpha Adducin|Alpha-adducin|ADDB
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Adducins are a family of cytoskeleton proteins encoded by three genes (alpha, beta and gamma). Adducin is a protein associated with the inner leaflet of the plasma membrane and is one of the proteins localized at the spectrin-Actin junction of the membrane skeleton. The cortical Actin cytoskeletal network is lost during apoptosis and Adducins are central in the cortical Actin network organization. Adducin alpha is a cytoskeletal protein involved with sodium-pump activity in the renal tubule and is associated with hypertension. The expression of Adducin alpha and Adducin gamma is ubiquitous in contrast to the restricted expression of Adducin beta . Adducin beta is expressed at high levels in brain and hematopoietic tissues, such as bone marrow in humans and spleen in mice.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ADD1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat