Anti-MIDN Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-0762R-A647
Nome del fornitore: Bioss
Specifiche
- Antibody type:Primary
- Antigen name:Midnolin
- Antigen symbol:MIDN
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:90007
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifiche
Informazioni su questo articolo
Midnolin
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MIDN
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat