Specifications
- Antibody type:Primary
- Antigen name:Calcium Channel, Voltage-dependent, L type, alpha 1C subunit
- Antigen symbol:CACNA1C
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:775
- Antigen synonyms:cardic dihydropyridine-sensitive|alpha-1 subunit|voltage-gated calcium channel alpha subunit Cav1.2|calcium channel|isoform 1|voltage-gated L-type calcium channel Cav1.2 alpha 1 subunit|L type|TS|MGC120730|CACN2|CACNL1A1|CCHL1A1|alpha 1C subunit|CACNA1C|cardic muscle|alpha 1 polypeptide|voltage-dependent L-type calcium channel alpha 1C subunit|splice variant 10*|DHPR|voltage-dependent|CaV1.2|CACH2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
This gene encodes an alpha-1 subunit of a voltage-dependent calcium channel. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization. The alpha-1 subunit consists of 24 transmembrane segments and forms the pore through which ions pass into the cell. The calcium channel consists of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. There are multiple isoforms of each of these proteins, either encoded by different genes or the result of alternative splicing of transcripts. The protein encoded by this gene binds to and is inhibited by dihydropyridine. Alternative splicing results in many transcript variants encoding different proteins. [provided by RefSeq]
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CACNA1C
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat