Anti-CXCL1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-10234R-A488
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Chemokine (C-X-C Motif) Ligand 1
- Antigen symbol:CXCL1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:14825
- Antigen synonyms:SCYB1|GROa|GRO1|MGSA-a|NAP-3|MGSA|FSP
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:70-107/107
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Has chemotactic activity for neutrophils. Contributes to neutrophil activation during inflammation (By similarity). Hematoregulatory chemokine, which, in vitro, suppresses hematopoietic progenitor cell proliferation. KC(5-72) shows a highly enhanced hematopoietic activity.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CXCL1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity: