- Antibody type:Primary
- Antigen name:Large tumor suppressor kinase 1 And 2
- Antigen symbol:LATS1/2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:26524, 9113
- Antigen synonyms:WARTS|Large tumor suppressor homolog 1|KPM|Large tumor suppressor homolog 2|Serine/threonine-protein kinase kpm|Serine/threonine-protein kinase LATS1|LATS2|LATS1|Kinase phosphorylated during mitosis protein|Serine/threonine-protein kinase LATS2|h-warts|WARTS protein kinase|Warts-like kinase
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:640-690/1088
- Purification:Purified by Protein A
- Pk:100 µl
This gene encodes a serine/threonine protein kinase belonging to the LATS tumor suppressor family. The protein localizes to centrosomes during interphase, and early and late metaphase. It interacts with the centrosomal proteins aurora-A and ajuba and is required for accumulation of gamma-tubulin and spindle formation at the onset of mitosis. It also interacts with a negative regulator of p53 and may function in a positive feedback loop with p53 that responds to cytoskeleton damage. Additionally, it can function as a co-repressor of androgen-responsive gene expression. [provided by RefSeq].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: LATS1/2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat