Order Entry
Italy
ContactUsLinkComponent
Anti-AMPK beta 1 + 2 Rabbit Polyclonal Antibody (Cy7®)
Anti-AMPK beta 1 + 2 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-3967R-CY7
Supplier:  Bioss
Anti-AMPK beta 1 + 2 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-3967R-CY7
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    5' AMP-activated protein kinase beta 1 And 2
  • Antigen symbol:
    AMPK beta 1 + 2
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy7®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    5565, 5564
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    220-270/272
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

PRKAB2 ans PRKAB1 are regulatory subunits of the AMP-activated protein kinase (AMPK). AMPK is a heterotrimer consisting of an alpha catalytic subunit, and non-catalytic beta and gamma subunits. AMPK is an important energy-sensing enzyme that monitors cellular energy status and plays a role in protecting cells from stresses that cause ATP depletion by switching off ATP-consuming biosynthetic pathways. AMPK is responsible for the regulation of fatty acid synthesis by phosphorylation of acetyl-CoA carboxylase (ACC). It also regulates cholesterol synthesis via phosphorylation and inactivation of hydroxymethylglutaryl-CoA reductase (HMGCR) and hormone-sensitive lipase. PRKAB2 may be a positive regulator of AMPK activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AMPK beta 1 + 2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat