Specifications
- Antibody type:Primary
- Antigen name:Tripartite Motif Containing 63
- Antigen symbol:TRIM63
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:84676
- Antigen synonyms:tripartite motif-containing 63|muscle specific ring finger protein 2|MURF1|IRF|tripartite motif-containing protein 63|SMRZ|RNF28|iris ring finger protein|striated muscle RING zinc finger protein|MURF2|E3 ubiquitin-protein ligase TRIM63|muscle-specific RING finger protein 1|ring finger protein 28
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:280-315/353
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
This gene encodes a member of the RING zinc finger protein family found in striated muscle and iris. The product of this gene is localized to the Z-line and M-line lattices of myofibrils, where titin's N-terminal and C-terminal regions respectively bind to the sarcomere. In vitro binding studies have shown that this protein also binds directly to titin near the region of titin containing kinase activity. Another member of this protein family binds to microtubules. Since these family members can form heterodimers, this suggests that these proteins may serve as a link between titin kinase and microtubule-dependent signal pathways in muscle. [provided by RefSeq].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TRIM63
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat