Order Entry
Italy
ContactUsLinkComponent
 
Anti-PD1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
  BOSSBS-1867R-HRP
 :  Bioss
Anti-PD1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
  BOSSBS-1867R-HRP
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Programmed cell death protein 1
  • Antigen symbol:
    PD1
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    5133
  • Antigen synonyms:
    PD-1|hPD-1|hPD-l|programmed cell death protein 1|CD279|protein PD-1|PD1|SLEB2
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    240-288/288
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Inhibitory cell surface receptor involved in the regulation of T-cell function during immunity and tolerance. Upon ligand binding, inhibits T-cell effector functions in an antigen-specific manner. Possible cell death inducer, in association with other factors.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: PD1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat