Order Entry
Italy
ContactUsLinkComponent
Anti-MMP17 Rabbit Polyclonal Antibody (Cy3®)
Anti-MMP17 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-1862R-CY3
Supplier:  Bioss
Anti-MMP17 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-1862R-CY3
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Matrix Metalloproteinase 17
  • Antigen symbol:
    MMP17
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy3®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    4326
  • Antigen synonyms:
    MT4-MMP
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

The matrix metalloproteinases (MMPs) are a family of at least eighteen secreted and membrane bound zincendopeptidases. Collectively, these enzymes can degrade all the components of the extracellular matrix, including fibrillar and non fibrillar collagens, fibronectin, laminin and basement membrane glycoproteins. In general, a signal peptide, a propeptide, and a catalytic domain containing the highly conserved zinc binding site characterizes the structure of the MMPs. In addition, fibronectin like repeats, a hinge region, and a C terminal hemopexin like domain allow categorization of MMPs into the collagenase, gelatinase, stomelysin and membrane type MMP subfamilies. All MMPs are synthesized as proenzymes, and most of them are secreted from the cells as proenzymes. Thus, the activation of these proenzymes is a critical step that leads to extracellular matrix breakdown. MMPs are considered to play an important role in wound healing, apoptosis, bone elongation, embryo development, uterine involution, angiogenesis and tissue remodeling, and in diseases such as multiple sclerosis, Alzheimer's, malignant gliomas, lupus, arthritis, periodontis, glumerulonephritis, atherosclerosis, tissue ulceration, and in cancer cell invasion and metastasis.MMP17 has been reported to be elevated in several tumor cell lines, and is constituitively produced by some normal cell lines. Treatment of cells with Concanavolin A or the phorbol ester TPA stimulates production of MMP17 in some cell types, and the enzyme can be recovered in cell lysates. Shed forms of MMP17 have also been reported.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MMP17
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat