Order Entry
Italy
ContactUsLinkComponent
Anti-CCKBR Rabbit Polyclonal Antibody (Cy7®)
Anti-CCKBR Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-1777R-CY7
Supplier:  Bioss
Anti-CCKBR Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-1777R-CY7
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cholecystokinin B Receptor
  • Antigen symbol:
    CCKBR
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy7®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    887
  • Antigen synonyms:
    CCK-B|CCK2R|GASR
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    300-350/447
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Receptor for gastrin and cholecystokinin. The CKK-B receptors occur throughout the central nervous system where they modulate anxiety, analgesia, arousal, and neuroleptic activity. This receptor mediates its action by association with G proteins that activate a phosphatidylinositol-calcium second messenger system. Isoform 2 is constitutively activated and may regulate cancer cell proliferation via a gastrin-independent mechanism.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCKBR
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat