Order Entry
Italy
ContactUsLinkComponent
 
Anti-LRRC6 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-13613R-A555
Supplier:  Bioss
Anti-LRRC6 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-13613R-A555
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Leucine Rich Repeat Containing 6
  • Antigen symbol:
    LRRC6
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 555
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Antigen synonyms:
    LRTP|TSLRP|CILD19
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

The leucine-rich repeat-containing protein 6 (LRRC6), also designated Leucine-rich testis-specific protein (LRTP), is a 466 amino acid protein that contains 3 LRR repeats and plays a role in spermatogenesis. The gene encoding LRRC6 maps to chromosome 8, which encodes approximately 800 genes. Translocation of portions of chromosome 8 with amplifications of the c-Myc gene are found in some leukemias and lymphomas, and typically associated with a poor prognosis. Chromosome 8 is also associated with Trisomy 8, Pfeiffer syndrome, congenital hypothyroidism and Waardenburg syndrome.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LRRC6
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat