Anti-MPP2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
BOSSBS-13689R-A647
: Bioss
- Antibody type:Primary
- Antigen name:Membrane Palmitoylated Protein 2
- Antigen symbol:MPP2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Homolog 2|Membrane Protein|DLG2|Discs Large|MAGUK P55 Subfamily Member 2|Palmitoylated 2|Protein MPP2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
MPP2 (Palmitoylated membrane protein 2) is a member of a family of membrane associated proteins termed MAGUKs (membrane associated guanylate kinase homologs). These proteins interact with the cytoskeleton and regulate cell proliferation, signaling pathways, and intracellular junctions. MPP2 contains a conserved sequence, called the SH3 (src homology 3) motif, found in several other proteins that associate with the cytoskeleton and are suspected to play important roles in signal transduction.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MPP2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat