Anti-WDR68 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-13682R-FITC
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:WD Repeat Domain 68
- Antigen symbol:WDR68
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Seven WD repeat protein of the AN11 family 1|DCAF7_HUMAN|WD repeat domain 68|Petunia|HAN11|AN11|WD repeat-containing protein 68|WD repeat protein An11 homolog|DDB1- and CUL4-associated factor 7|WDR68.|SWAN 1|Dcaf7|WD repeat-containing protein An11 homolog|Human anthocyanin
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Involved in craniofacial development. Acts upstream of the EDN1 pathway and is required for formation of the upper jaw equivalent, the palatoquadrate. The activity required for EDN1 pathway function differs between the first and second arches (By similarity). Associates with DIAPH1 and controls GLI1 transcriptional activity. Could be involved in normal and disease skin development. May function as a substrate receptor for CUL4-DDB1 E3 ubiquitin-protein ligase complex.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: WDR68
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat