Anti-GPATCH2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-13499R-A647
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:G Patch Domain Containing 2
- Antigen symbol:GPATCH2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rabbit,Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:GPATC2|CT110|PPP1R30
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
GPATCH2
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GPATCH2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit