Order Entry
Italy
ContactUsLinkComponent
Anti-GAB4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-GAB4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-13256R-FITC
Supplier:  Bioss
Anti-GAB4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-13256R-FITC
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    GRB2-associated binding protein 4
  • Antigen symbol:
    GAB4
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    128954
  • Antigen synonyms:
    GRB2-Associated Binder 4|Growth Factor Receptor Bound Protein 2-Associated Protein 4|GRB2-Associated Binding Protein Family|Member 4|GRB2-Associated-Binding Protein 2-Like|GAB2-Like|GRB2-Associated Binder 2-Like
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

The Gab family of adaptor proteins function as molecular scaffolds that mediate protein recruit-ment to RTKs. Cytokine/growth factor triggering of protein tyrosine kinase receptors (RTKs) initiates signaling cascades that progress to the nucleus where signals for activation, proliferation and differentiation occur. This scaffolding mechanism represents a critical link in cytokine/growth factor signaling routes. Gab 1-4 contain Pleckstrin homology and potential binding sites for SH2 and SH3 domain-containing proteins. The recruitment of signaling partners to Gab family members is phosphorylation-dependent. Insulin receptor and EGF receptor signaling are among the cascades that rely on Gab family members to elicit a nuclear response to an extracellular stimulus. Gab 4 (GRB2-associated-binding protein 4), also designated GRB2-associated-binding protein 2-like (Gab 2-like), is a 574 amino acid protein that shares 62% sequence similarity with Gab 2 and contains one Pleckstrin homology domain.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GAB4
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat