Order Entry
Italy
ContactUsLinkComponent
Anti-GDF8 Goat Polyclonal Antibody (Alexa Fluor® 350)
Anti-GDF8 Goat Polyclonal Antibody (Alexa Fluor® 350)
  BOSSBS-1288G-A350
 :  Bioss
Anti-GDF8 Goat Polyclonal Antibody (Alexa Fluor® 350)
  BOSSBS-1288G-A350
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Growth Differentiation Factor 8
  • Antigen symbol:
    GDF8
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 350
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Antigen synonyms:
    Growth/Differentiation Factor 8|Growth differentiation factor 8|myostatin.|MSTN|growth differntiation factor 8|GDF 8
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Myostatin (GDF8)is expressed uniquely in human skeletal muscle as a 12 kDa mature glycoprotein consisting of 113 amino acid residues and secreted into plasma. Myostatin is a member of the transforming growth factor beta superfamily of secreted growth and differentiation factors that is essential for proper regulation of skeletal muscle mass. Studies have shown that myostatin could play an important role in cardiac development and physiology.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GDF8
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat