- Antibody type:Primary
- Antigen name:Chondroitin sulfate N-acetylgalactosaminyltransferase 1
- Antigen symbol:CSGALNACT1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:55790
- Antigen synonyms:beta4GalNAcT|ChGn|CSGalNAcT-1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
The chondroitin N-acetylgalactosaminyltransferase family includes Beta-1,4-GalNAc-T, Beta-1,4-GalNAc-T2, Beta-1,4-GalNAc-T3 and Beta-1,4-GalNAc-T4. The Beta-1,4-GalNAc-T protein consists of a short N-terminal residue, a transmembrane region and a long C-terminal residue, which includes a catalytic domain and localizes to the Golgi apparatus. Beta-1,4-GalNAc-T utilizes simple ganglioside GM3 as a substrate for more complex gangliosides GM2, GM1 and GD1a. Beta-1,4-GalNAc-T is expressed in normal brain tissues and in various malignant transformed cells, such as malignant melanoma, neuroblastoma and adult T cell leukemia. Mice lacking the Beta-1,4-GalNAc-T protein develop significant and progressive behavioral neuropathies, including deficits in reflexes, strength, coordination and balance. Beta-1,4-GalNAc-T is a potential molecular marker for detecting melanoma cells and monitoring tumor progression.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CSGALNACT1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity: