- Antibody type:Primary
- Antigen name:Attractin
- Antigen symbol:ATRN
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:8455
- Antigen synonyms:DPPT-L|MGCA
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Mahogany (MG), originally identified as a protein involved in pigmentation, acts in conjunction with melanocortin receptors to suppress diet-induced obesity. Mahogany contains a single transmembrane domain, and it is expressed in a broad range of tissues, including the hypothalamus and pigment cells. Mutations within the mahogany gene were shown to rescue agouti-lethal-yellow mutant mice from obesity. The extracellular domain of mouse mahogany is the ortholog of the human protein attractin. Attractin (also designated DPPT-L) is a human serum glycoprotein and is a member of the CUB family of cell adhesion and guidance proteins. Attractin is expressed on activated T cells and is released from the cells 48 to 72 hours after activation.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: ATRN
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat