Order Entry
Italy
ContactUsLinkComponent
Anti-NAC1/BTBD14B Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anti-NAC1/BTBD14B Rabbit Polyclonal Antibody (Alexa Fluor® 647)
  BOSSBS-12247R-A647
 :  Bioss
Anti-NAC1/BTBD14B Rabbit Polyclonal Antibody (Alexa Fluor® 647)
  BOSSBS-12247R-A647
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen symbol:
    NAC1/BTBD14B
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 647
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    112939
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

NAC1 is an N-terminal homodimerization domain that contains multiple copies of kelch repeats and/or C2H2-type zinc fingers. Proteins that contain BTB domains are thought to be involved in transcriptional regulation via control of chromatin structure and function. BTBD14B (BTB/POZ domain-containing protein 14B), also known as NACC1 (nucleus accumbens associated 1), BEND8 or NAC1, is a 527 amino acid protein that localizes to both the nucleus and the cytoplasm and contains one BTB (POZ) domain. Existing as a homooligomer that interacts with HDAC3 and HDAC4, BTBD14B functions as a transcriptional repressor that influences the transcriptional activity of CRIF1 and is required for proteasome recruitment to the nucleus and cytoplasm in dendritic spines. BTBD14B is overexpressed in multiple carcinomas, suggesting a role in tumor development and metastasis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NAC1/BTBD14B
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat