Order Entry
Italy
ContactUsLinkComponent
Anti-ATP6V1B2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anti-ATP6V1B2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-12549R-A488
Supplier:  Bioss
Anti-ATP6V1B2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-12549R-A488
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B2
  • Antigen symbol:
    ATP6V1B2
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 488
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    526
  • Antigen synonyms:
    ATP6B1B2|HO57|VATB|Vma2|ATP6B2|VPP3
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Vacuolar-type H+-ATPase (V-ATPase) is a multisubunit enzyme responsible for acidification of eukaryotic intracellular organelles. V-ATPases pump protons against an electrochemical gradient, while F-ATPases reverse the process, thereby synthesizing ATP. A peripheral V1 domain, which is responsible for ATP hydrolysis, and a integral V0 domain, which is responsible for proton translocation, compose V-ATPase. Nine subunits (A–H) make up the V1 domain and five subunits (a, d, c, c' and c") make up the V0 domain. Like F-ATPase, V-ATPase most likely operates through a rotary mechanism. The V-ATPase V1 B subunit exists as two isoforms. In the inner ear, the V-ATPase B1 isoform functions in proton secretion and is required to maintain proper endolymph pH and normal auditory function. The gene encoding the human V-ATPase B1 isoform maps to chromosome 2cen-q13. Mutations in this gene cause distal renal tubular acidosis associated with sensorineural deafness. The V-ATPase B2 isoform is expressed in kidney and is the only B isoform expressed in osteoclasts. The gene encoding the human V-ATPase B2 isoform maps to chromosome 8p22-p21.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ATP6V1B2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat