Specifications
- Antibody type:Primary
- Antigen name:Striatin, Calmodulin Binding Protein 3
- Antigen symbol:STRN3
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:STRN3 |Cell-cycle autoantigen SG2NA |SG2NA |S/G2 antigen |GS2NA
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Striatin, SG2NA, and zinedin, the three mammalian members of the striatin family, are multimodular, WD repeat and calmodulin-binding proteins. Zinedin and SG2NA share with striatin identical protein-protein interaction domains and the same overall domain structure. All three proteins are both cytosolic and membrane-bound and bind calmodulin in the presence of calcium. Striatin is a neuronal, intracellular protein strictly expressed in the somato-dendritic compartment, including spines and subsets of neurons, and is considered as a marker of neuronal polarity. Downregulation of striatin, which is expressed in a few subsets of neurons, impairs the growth of dendrites as well as rat locomotor activity. Zinedin is mainly expressed in the central nervous system, whereas SG2NA is mainly expressed in the brain and muscle.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: STRN3
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat