Order Entry
Italy
ContactUsLinkComponent
Anti-GABARAP Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anti-GABARAP Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-12082R-A488
Supplier:  Bioss
Anti-GABARAP Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-12082R-A488
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Gamma-aminobutyric acid receptor-associated protein
  • Antigen symbol:
    GABARAP
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 488
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Antigen synonyms:
    GABARAP-a|MM46|GABA(A) receptor-associated protein|FLC3B|ATG8A|Gamma-aminobutyric acid receptor-associated protein
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

In the central nervous system GABA functions as the main inhibitory transmitter by increasing a Cl-conductance that inhibits neuronal firing. GABA has been shown to activate both ionotropic (GABAA ) and metabotropic (GABAB ) receptors as well as a third class of receptors called GABAC. In addition to GABA receptors, several proteins have been identified as regulators of GABA function, including GAD65, GAD67, GABA transporters and GABARAP (GABAA receptor-associated protein). GABARAP associates with GABAA Rg2 to link GABAA receptors to the cytoskeleton. The GABARAP protein sequence is similar to light chain-3 of microtubule-associated proteins (MAPs) suggesting that it may be a type of MAP or a component of a MAP complex.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GABARAP
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat