Anti-RTN4R Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-0129R-A350
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Reticulon 4 Receptor
- Antigen symbol:RTN4R
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:65079
- Antigen synonyms:NGR|NGR1|NOGOR
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:140-190/473
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Receptor for RTN4, OMG and MAG. Mediates axonal growth inhibition and may play a role in regulating axonal regeneration and plasticity in the adult central nervous system (By similarity). Acts in conjunction with RTN4 and LIGO1 in regulating neuronal precursor cell motility during cortical development.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RTN4R
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat