Order Entry
Italy
ContactUsLinkComponent
Zinc acetate dihydrate 98.0-102.0%, Technical Grade, J.T.Baker®
Zinc acetate dihydrate 98.0-102.0%, Technical Grade, J.T.Baker®
Catalog # 4304-R
Supplier:  Avantor
CAS Number:  
undefined
Zinc acetate dihydrate 98.0-102.0%, Technical Grade, J.T.Baker®
Catalog # 4304-R
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Fibre drum
  • Pk:
    250 lb
  • Seq:
    7215

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    98.0 - 102.0 %
  • Assay ((CH₃COO)₂Zn·2H₂O):
    98.0 - 102.0 %
  • Insoluble Matter:
    ≤0.01 %
  • pH of 5% Solution at 25°C:
    6.0 - 7.0
  • Lead (Pb):
    ≤0.005 %

Specification Test Results

About this item

 :
 :219.51 g/mol
UN:3077
ADR:9,III
CAS Number:5970-45-6
Formula:(H₃CCOO)₂Zn·2H₂O
MDL Number:MFCD00066961