Silica gel, BAKER ANALYZED® 0.063 - 0.200 mm for chromatography, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:63231-67-4
:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232
:60.08 g/mol
Storage Temperature:Ambient
Specification Test Results
- Alcohol-Ether Soluble Substances:max. 0.05 %
- Loss on Drying:max. 6.0 %
Specification Test Results
Frequently Bought Together