Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Potassium chloride
Potassium chloride ACS
Potassium chloride ACS
Potassium chloride ACS
Catalog # HACH76401
Supplier:
Hach
CAS Number:
Expand image
Potassium chloride ACS
Catalog # HACH76401
Supplier:
Hach
CAS Number:
Specifications
Pack type:
Bottle
Pk:
454 g
Seq:
3635
Specifications
About this item
Product Details
:
:
74.55 g/mol
CAS Number:
7447-40-7
Formula:
KCl
MDL Number:
MFCD00011360
Product Details