Anti-IgG, C3 IgG Polyclonal Antibody
ICNA08646381
- Antibody type:Primary
- Antigen name:IgG, C3
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Cat
- Seq:02
- Format:Antiserum
- Form:Lyophilized
- Storage temperature:+2 °C to +8 °C
- Shipping temperature:RT
- Tested applications:Autoimmune hemolytic anemia screening
- Size:200 µl
- Pk:2 mL
Type: 20269687
Antigen: Primary
Clonality: IgG, C3
Clone: Polyclonal
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Cat