Order Entry
Italy
ContactUsLinkComponent
Anti-MMP-16 Rabbit Polyclonal Antibody [clone: T583]
Anti-MMP-16 Rabbit Polyclonal Antibody [clone: T583]
Catalog # BWRLBS1234
Anti-MMP-16 Rabbit Polyclonal Antibody [clone: T583]
Catalog # BWRLBS1234

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    MMP-16
  • Clonality:
    Polyclonal
  • Clone:
    T583
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to amino acids 550-600 of Human MMP-16.

MMP-16 (T583) pAb detects endogenous levels of MMP-16 protein.

Type: Primary
Antigen: MMP-16
Clonality: Polyclonal
Clone: T583
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat