Anti-Ku-70 Rabbit Polyclonal Antibody [clone: R586]
BWRLBS1212
- Antibody type:Primary
- Antigen symbol:Ku-70
- Clonality:Polyclonal
- Clone:R586
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 560-600 of Human Ku-70.
Ku70 (R586) pAb detects endogenous levels of Ku70 protein.
Type: Primary
Antigen: Ku-70
Clonality: Polyclonal
Clone: R586
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat