Anti-Cleaved-MMP-23 Rabbit Polyclonal Antibody [clone: Y79]
Catalog # BWRLBS7054
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Cleaved-MMP-23
- Clonality:Polyclonal
- Clone:Y79
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 51-100 of Human MMP-23.
Cleaved-MMP-23 (Y79) pAb detects endogenous levels of Cleaved-MMP-23 (37 kDa) protein.
Type: Primary
Antigen: Cleaved-MMP-23
Clonality: Polyclonal
Clone: Y79
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat