Anti-Cathepsin L2 Rabbit Polyclonal Antibody [clone: L114]
Catalog # BWRLBS7023
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Cathepsin L2
- Clonality:Polyclonal
- Clone:L114
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 91-140 of Human Cathepsin L2.
Cathepsin L2 (L114) pAb detects endogenous levels of Cathepsin L2 protein.
Type: Primary
Antigen: Cathepsin L2
Clonality: Polyclonal
Clone: L114
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat