Anti-EIF4B Rabbit Polyclonal Antibody
BWRLBS7572
- Antibody type:Primary
- Antigen symbol:EIF4B
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Recombinant full length Human EIF4B.
EIF4B pAb detects endogenous levels of EIF4B protein.
Type: Primary
Antigen: EIF4B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat